CAS RN 88145-89-5
CAS RN 88145-89-5
IUPAC Name:
6-bromo-1,2,3,4-tetrahydroquinazoline-2,4-dione
Synonyms:
6-bromo-1,3-dihydroquinazoline-2,4-dione
Molecular Weight (g/mol):
241.04
Molecular Formula:
C8H5BrN2O2
InChi Key:
JZDVFUAHGLJVQG-UHFFFAOYSA-N
SMILES:
BrC1=CC=C2NC(=O)NC(=O)C2=C1
1
–
1
of
1
results
1
6-Bromoquinazoline-2,4(1H,3H)-dione, 97%
CAS: 88145-89-5 Molecular Formula: C8H5BrN2O2 Molecular Weight (g/mol): 241.044 MDL Number: MFCD00462868 InChI Key: JZDVFUAHGLJVQG-UHFFFAOYSA-N Synonym: 6-bromoquinazoline-2,4 1h,3h-dione,6-bromo-2,4 1h,3h-quinazolinedione,6-bromoquinazoline-2,4-dione,6-bromoquinazoline-2,4-diol,2,4 1h,3h-quinazolinedione, 6-bromo,6-bromo-1,3-dihydroquinazoline-2,4-dione,6-bromo-1,2,3,4-tetrahydroquinazoline-2,4-dione,bromoquinazolinedione,pubchem20961,acmc-209qre PubChem CID: 617686 IUPAC Name: 6-bromo-1H-quinazoline-2,4-dione SMILES: C1=CC2=C(C=C1Br)C(=O)NC(=O)N2