CAS RN 885267-95-8
IUPAC Name:
3',5'-dichloro-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms:
3',5'-dichloro-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol):
321.6
Molecular Formula:
C12H7Cl3O2S
InChi Key:
KBMKQHVXGMOKKI-UHFFFAOYSA-N
SMILES:
ClC1=CC(=CC(Cl)=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
3',5'-Dichlorobiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific™
CAS: 885267-95-8 Molecular Formula: C12H7Cl3O2S Molecular Weight (g/mol): 321.61 InChI Key: KBMKQHVXGMOKKI-UHFFFAOYSA-N Synonym: 4-3,5-dichlorophenyl benzenesulfonyl chloride,3',5'-dichloro-1,1'-biphenyl-4-sulfonyl chloride,4-3,5-dichlorophenyl benzenesulphonyl chloride,3',5'-dichlorobiphenyl-4-sulfonyl chloride,4-3,5-dichlorophenyl phenyl sulfonyl chloride PubChem CID: 3771398 IUPAC Name: 4-(3,5-dichlorophenyl)benzenesulfonyl chloride SMILES: C1=CC(=CC=C1C2=CC(=CC(=C2)Cl)Cl)S(=O)(=O)Cl