CAS RN 885325-71-3
CAS RN 885325-71-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MK-8745, MedChemExpress
MedChemExpress MK-8745 is an aurora A kinase inhibitor with an IC50 of 0.6 nM.