accessibility menu, dialog, popup

UserName

CAS RN 886500-41-0

FNFCl

CAS RN 886500-41-0

IUPAC Name: 4-chloro-2,6-difluorobenzonitrile
Synonyms: 4-chloro-2,6-difluorobenzonitrile
Molecular Weight (g/mol): 173.55
Molecular Formula: C7H2ClF2N
InChi Key: ABNYHMNNQYZWIM-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(F)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results