accessibility menu, dialog, popup

UserName

H3COOFF

CAS RN 887267-04-1

IUPAC Name: 3,6-difluoro-2-methoxybenzaldehyde
Synonyms: 3,6-difluoro-2-methoxybenzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: CRIGYCXILOWIKP-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(F)=C1C=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results