accessibility menu, dialog, popup

UserName

CAS RN 89167-19-1

NIBr

CAS RN 89167-19-1

IUPAC Name: 3-bromo-4-iodopyridine
Synonyms: 3-bromo-4-iodopyridine
Molecular Weight (g/mol): 283.89
Molecular Formula: C5H3BrIN
InChi Key: LKDNPXCBLVOBRP-UHFFFAOYSA-N
SMILES: BrC1=C(I)C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results