accessibility menu, dialog, popup

UserName

CH3NOSOOCl

CAS RN 892948-94-6

IUPAC Name: 4-methyl-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride
Synonyms: 4-methyl-2,3-dihydro-1,4-benzoxazine-6-sulfonyl chloride
Molecular Weight (g/mol): 247.69
Molecular Formula: C9H10ClNO3S
InChi Key: LHMOALYDXXCXLZ-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1C=C(C=C2)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results