accessibility menu, dialog, popup

UserName

CAS RN 89364-31-8

OOOH

CAS RN 89364-31-8

IUPAC Name: oxolane-3-carboxylic acid
Synonyms: oxolane-3-carboxylic acid
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: BOTREHHXSQGWTR-UHFFFAOYNA-N
SMILES: OC(=O)C1CCOC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results