CAS RN 89396-94-1
CAS RN 89396-94-1
IUPAC Name:
hydrogen (4S)-3-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-1-methyl-2-oxoimidazolidine-4-carboxylic acid chloride
Synonyms:
hydrogen imidapril chloride
Molecular Weight (g/mol):
441.91
Molecular Formula:
C20H28ClN3O6
InChi Key:
LSLQGMMMRMDXHN-GEUPQXMHSA-N
SMILES:
[H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1[C@@H](CN(C)C1=O)C(O)=O
1
–
1
of
1
results
1
Imidapril hydrochloride, MedChemExpress
MedChemExpress Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. Imidapril hydrochloride suppresses the conversion of angiotensin I to angiotensin II and thereby reduces total peripheral resistance and systemic blood pressure. Imidapril hydrochloride can be used for hypertension, type 1 diabetic, nephropathy and chronic heart failure research.