CAS RN 894779-67-0
IUPAC Name:
1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)methanamine
Synonyms:
1-(5-methyl-1-phenylpyrazol-4-yl)methanamine
Molecular Weight (g/mol):
187.25
Molecular Formula:
C11H13N3
InChi Key:
GRLXUIRJMGNTQO-UHFFFAOYSA-N
SMILES:
CC1=C(CN)C=NN1C1=CC=CC=C1
1
–
1
of
1
results
1
5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 95%, Thermo Scientific™
CAS: 894779-67-0 Molecular Formula: C11H13N3 Molecular Weight (g/mol): 187.24 InChI Key: GRLXUIRJMGNTQO-UHFFFAOYSA-N Synonym: 5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine,1h-pyrazole-4-methanamine, 5-methyl-1-phenyl,5-methyl-1-phenylpyrazol-4-yl methanamine,5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine PubChem CID: 28472963 IUPAC Name: (5-methyl-1-phenylpyrazol-4-yl)methanamine SMILES: CC1=C(C=NN1C2=CC=CC=C2)CN