CAS RN 89490-05-1
CAS RN 89490-05-1
IUPAC Name:
(cyclohex-1-en-1-yl)boronic acid
Synonyms:
cyclohex-1-en-1-ylboronic acid
Molecular Weight (g/mol):
125.96
Molecular Formula:
C6H11BO2
InChi Key:
XZWQKJXJNKYMAP-UHFFFAOYSA-N
SMILES:
OB(O)C1=CCCCC1
1
–
2
of
2
results
2
Cyclohexene-1-boronic acid, 97%, Thermo Scientific™
CAS: 89490-05-1 Molecular Formula: C6H11BO2 Molecular Weight (g/mol): 125.96 MDL Number: MFCD02179497 InChI Key: XZWQKJXJNKYMAP-UHFFFAOYSA-N Synonym: cyclohex-1-en-1-ylboronic acid,1-cyclohexenylboronic acid,1-cyclohexen-1-yl-boronic acid,cyclohexenylboronic acid,cyclohexen-1-yl-boronic acid,1-cyclohexen-1-ylboronic acid,cyclohexene-1-boronic acid,1-boronocyclohex-1-ene,cyclohex-1-enylboronic acid,cyclohexen-1-yl boronic acid PubChem CID: 3862902 IUPAC Name: cyclohexen-1-ylboronic acid SMILES: OB(O)C1=CCCCC1