CAS RN 896736-21-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ramelteon Metabolite M-II (mixture of R and S at the hydroxy position), TRC
CAS: 896736-21-3 Molecular Formula: C16 H21 N O3 Molecular Weight (g/mol): 275.34 Synonym: Ramelteon Metabolite M-Ii IUPAC Name: 2-hydroxy-N-[2-(2,6,7,8-tetrahydro-1H-cyclopenta[e][1]benzofuran-8-yl)ethyl]propanamide SMILES: CC(O)C(=O)NCCC1CCc2ccc3OCCc3c12