accessibility menu, dialog, popup

UserName

CAS RN 89809-64-3

NClN

CAS RN 89809-64-3

IUPAC Name: 5-chloropyridine-2-carbonitrile
Synonyms: 5-chloropyridine-2-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: WTHODOKFSYPTKA-UHFFFAOYSA-N
SMILES: ClC1=CN=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results