accessibility menu, dialog, popup

UserName

CAS RN 902837-30-3

CH3ClOHN

CAS RN 902837-30-3

IUPAC Name: 3-(2-chloro-5-methylphenoxy)piperidine
Synonyms: 3-(2-chloro-5-methylphenoxy)piperidine
Molecular Weight (g/mol): 225.72
Molecular Formula: C12H16ClNO
InChi Key: GRLVAJJXUNOVRA-UHFFFAOYNA-N
SMILES: CC1=CC(OC2CCCNC2)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results