accessibility menu, dialog, popup

UserName

CAS RN 902837-33-6

H3CNHOONH

CAS RN 902837-33-6

IUPAC Name: 2-(piperidin-3-yloxy)-N-propylacetamide
Synonyms: 2-(piperidin-3-yloxy)-N-propylacetamide
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: HIPDESKOLOAVFF-UHFFFAOYNA-N
SMILES: CCCNC(=O)COC1CCCNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results