accessibility menu, dialog, popup

UserName

OH(S)NH(S)HO

CAS RN 90365-74-5

IUPAC Name: (3S,4S)-1-benzyl-3,4-dihydroxypyrrolidin-1-ium
Synonyms: (3S,4S)-1-benzyl-3,4-dihydroxypyrrolidin-1-ium
Molecular Weight (g/mol): 194.25
Molecular Formula: C11H16NO2
InChi Key: QJRIUWQPJVPYSO-QWRGUYRKSA-O
SMILES: O[C@H]1C[NH+](CC2=CC=CC=C2)C[C@@H]1O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)