accessibility menu, dialog, popup

UserName

CAS RN 90381-08-1

FOFFF

CAS RN 90381-08-1

IUPAC Name: 5-fluoro-2-(trifluoromethyl)benzaldehyde
Synonyms: 5-fluoro-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: MITVIZWTPVYRDO-UHFFFAOYSA-N
SMILES: FC1=CC=C(C(C=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results