accessibility menu, dialog, popup

UserName

CAS RN 905281-76-7

NOHNNNOH(E/Z)

CAS RN 905281-76-7

IUPAC Name: 2-{4-[1-(hydroxyimino)-2,3-dihydro-1H-inden-5-yl]-3-(pyridin-4-yl)-1H-pyrazol-1-yl}ethan-1-ol
Synonyms: 2-{4-[1-(hydroxyimino)-2,3-dihydroinden-5-yl]-3-(pyridin-4-yl)pyrazol-1-yl}ethanol
Molecular Weight (g/mol): 334.38
Molecular Formula: C19H18N4O2
InChi Key: DEZZLWQELQORIU-UHFFFAOYSA-N
SMILES: OCCN1C=C(C(=N1)C1=CC=NC=C1)C1=CC2=C(C=C1)C(CC2)=NO
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (4)
  • (1)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)