accessibility menu, dialog, popup

UserName

HNH2NNON(E/Z)

CAS RN 90993-49-0

IUPAC Name: 1-(1,2-dihydropyrimidin-2-ylidene)-1-nitrosomethanamine
Synonyms: 1-nitroso-1-(1H-pyrimidin-2-ylidene)methanamine
Molecular Weight (g/mol): 138.13
Molecular Formula: C5H6N4O
InChi Key: RYYZBKPHKBOVSQ-UHFFFAOYSA-N
SMILES: NC(N=O)=C1NC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results