accessibility menu, dialog, popup

UserName

H2N

CAS RN 91-59-8

IUPAC Name: naphthalen-2-amine
Synonyms: β naphthylamine 2-aminonaftalen 2-naphthylamine and its salts 2-naphthylamine (substance) β-naftyloamina 2-naphthylamine mustard BNA β-naftylamin β-naftalamin fast scarlet base B
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: JBIJLHTVPXGSAM-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results