CAS RN 911434-05-4
IUPAC Name:
5-bromo-2-methyl-3-nitropyridine
Synonyms:
5-bromo-2-methyl-3-nitropyridine
Molecular Weight (g/mol):
217.02
Molecular Formula:
C6H5BrN2O2
InChi Key:
FZZLWWNOYMHSIS-UHFFFAOYSA-N
SMILES:
CC1=NC=C(Br)C=C1[N+]([O-])=O
1
–
1
of
1
results
1
5-Bromo-2-methyl-3-nitropyridine, 96%, Thermo Scientific Chemicals
CAS: 911434-05-4 Molecular Formula: C6H5BrN2O2 Molecular Weight (g/mol): 217.022 MDL Number: MFCD09031419 InChI Key: FZZLWWNOYMHSIS-UHFFFAOYSA-N Synonym: 5-bromo-3-nitropicoline,5-bromo-3-nitro-2-picoline,5-bromo-2-methyl-3-nitro-pyridine,pyridine, 5-bromo-2-methyl-3-nitro,pubchem13809,ksc496e9t,bromo-2-methyl-3-nitropyridine PubChem CID: 25067266 IUPAC Name: 5-bromo-2-methyl-3-nitropyridine SMILES: CC1=NC=C(C=C1[N+](=O)[O-])Br