CAS RN 91170-93-3
IUPAC Name:
3-chloro-4-fluorobenzene-1-sulfonyl chloride
Synonyms:
3-chloro-4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol):
229.05
Molecular Formula:
C6H3Cl2FO2S
InChi Key:
DXFXNSNBZNELII-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1Cl)S(Cl)(=O)=O
1
–
1
of
1
results
1
3-Chloro-4-fluorobenzenesulfonyl chloride, 98%
CAS: 91170-93-3 Molecular Formula: C6H3Cl2FO2S Molecular Weight (g/mol): 229.046 MDL Number: MFCD00041444 InChI Key: DXFXNSNBZNELII-UHFFFAOYSA-N Synonym: 3-chloro-4-fluorobenzene-1-sulfonyl chloride,3-chloro-4-fluorobenzenesulphonyl chloride,3-chloro-4-fluorobenzenesulfonylchloride,benzenesulfonyl chloride, 3-chloro-4-fluoro,3-chloro-4-fluorophenylsulfonyl chloride,3-chloro-4-fluoro-benzenesulfonyl chloride,benzenesulfonyl chloride, 3-chloro-4-fluoro-9ci,pubchem5093,acmc-209r8d,ksc488g2p PubChem CID: 145222 IUPAC Name: 3-chloro-4-fluorobenzenesulfonyl chloride SMILES: C1=CC(=C(C=C1S(=O)(=O)Cl)Cl)F