accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3C

CAS RN 911708-01-5

IUPAC Name: 2-(4-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 294.2
Molecular Formula: C19H23BO2
InChi Key: FPLGWDLJBAAWHO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CC2=CC=CC=C2)C=C1
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results