accessibility menu, dialog, popup

UserName

CAS RN 914311-50-5

BrNHNH2N

CAS RN 914311-50-5

IUPAC Name: 4-bromo-1H-indazol-3-amine
Synonyms: 4-bromo-1H-indazol-3-amine
Molecular Weight (g/mol): 212.05
Molecular Formula: C7H6BrN3
InChi Key: IXXXOIXGQFPLIH-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC=CC(Br)=C12
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)