accessibility menu, dialog, popup

UserName

OFFFBrNH2

CAS RN 914636-35-4

IUPAC Name: 3-bromo-5-(trifluoromethoxy)aniline
Synonyms: 3-bromo-5-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 256.02
Molecular Formula: C7H5BrF3NO
InChi Key: GJUHRBPRJGNGFH-UHFFFAOYSA-N
SMILES: NC1=CC(OC(F)(F)F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results