accessibility menu, dialog, popup

UserName

CAS RN 915707-70-9

CH3HNNBr

CAS RN 915707-70-9

IUPAC Name: [(5-bromopyridin-2-yl)methyl](methyl)amine
Synonyms: [(5-bromopyridin-2-yl)methyl](methyl)amine
Molecular Weight (g/mol): 201.07
Molecular Formula: C7H9BrN2
InChi Key: DCQLMECRAAGXSD-UHFFFAOYSA-N
SMILES: CNCC1=NC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results