accessibility menu, dialog, popup

UserName

CH3OFFFNH2

CAS RN 916420-96-7

IUPAC Name: 1-[3-methoxy-5-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[3-methoxy-5-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 205.18
Molecular Formula: C9H10F3NO
InChi Key: WNCDSBLUNYDXQO-UHFFFAOYSA-N
SMILES: COC1=CC(CN)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results