accessibility menu, dialog, popup

UserName

CAS RN 916766-82-0

CH3NNH2N

CAS RN 916766-82-0

IUPAC Name: 4-(1-methyl-1H-pyrazol-3-yl)aniline
Synonyms: 4-(1-methylpyrazol-3-yl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: WEFJHPBVOKVCLZ-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results