accessibility menu, dialog, popup

UserName

CH3NHNF

CAS RN 916792-26-2

IUPAC Name: 5-(4-fluorophenyl)-4-methyl-1H-pyrazole
Synonyms: 3-(4-fluorophenyl)-4-methyl-2H-pyrazole
Molecular Weight (g/mol): 176.19
Molecular Formula: C10H9FN2
InChi Key: DSDCWXOENGTWKJ-UHFFFAOYSA-N
SMILES: CC1=C(NN=C1)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results