CAS RN 917562-33-5
CAS RN 917562-33-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ JT010
Potent and selective TRPA1 channel activator (EC50 = 65 nM)
2