CAS RN 91993-31-6
CAS RN 91993-31-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oxynitrox∣r S100, free radical, 99+%, average M.W. ca 2,250
CAS: 91993-31-6 Synonym: PIPO