accessibility menu, dialog, popup

UserName

CAS RN 92-95-5

NCO

CAS RN 92-95-5

IUPAC Name: 4-isocyanato-1,1'-biphenyl
Synonyms: p-xenylcarbimide
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: WIRPZDICFIIBRF-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)