CAS RN 920759-93-9
CAS RN 920759-93-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-chloro-3-methylbenzoate, TRC
CAS: 920759-93-9 Molecular Formula: C9H9ClO2 Molecular Weight (g/mol): 184.62 IUPAC Name: methyl 2-chloro-3-methylbenzoate SMILES: COC(=O)c1cccc(C)c1Cl