CAS RN 921938-81-0
IUPAC Name:
{4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-7-yl}methanol
Synonyms:
{4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazin-7-yl}methanol
Molecular Weight (g/mol):
180.21
Molecular Formula:
C9H12N2O2
InChi Key:
PPBFQXGUORHMBA-UHFFFAOYSA-N
SMILES:
CN1CCOC2=C1N=CC(CO)=C2
1
–
1
of
1
results
1
(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methanol, 97%, Thermo Scientific™
CAS: 921938-81-0 Molecular Formula: C9H12N2O2 Molecular Weight (g/mol): 180.207 MDL Number: MFCD09817507 InChI Key: PPBFQXGUORHMBA-UHFFFAOYSA-N Synonym: 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol,4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol,4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol,2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl PubChem CID: 24229648 IUPAC Name: (4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol SMILES: CN1CCOC2=C1N=CC(=C2)CO