accessibility menu, dialog, popup

UserName

CAS RN 921939-11-9

CH3HNNN

CAS RN 921939-11-9

IUPAC Name: methyl[(2-phenylpyrimidin-5-yl)methyl]amine
Synonyms: methyl[(2-phenylpyrimidin-5-yl)methyl]amine
Molecular Weight (g/mol): 199.26
Molecular Formula: C12H13N3
InChi Key: BEURLXDVKSQEIQ-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results