CAS RN 92339-11-2
CAS RN 92339-11-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
No Refinements
1
–
0
of
0
results
1
Iodixanol, TRC
CAS: 92339-11-2 Molecular Formula: C35 H44 I6 N6 O15 Molecular Weight (g/mol): 1550.18 Synonym: 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(acetylimino)]bis[N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide,2-5410-3A,Iodixanol,OptiPrep,Visipaque,Visipaque 270 IUPAC Name: 5-[acetyl-[3-[N-acetyl-3,5-bis(2,3-dihydroxypropylcarbamoyl)-2,4,6-triiodoanilino]-2-hydroxypropyl]amino]-1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide SMILES: CC(=O)N(CC(O)CN(C(=O)C)c1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I)c2c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c2I