CAS RN 92739-63-4
CAS RN 92739-63-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
meso-Tetra (N-Methyl-4-pyridyl) Porphine Tetrachloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
meso-Tetra (N-Methyl-4-pyridyl) Porphine Tetrachloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
meso-Tetra (N-Methyl-4-pyridyl) Porphine Tetrachloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.