accessibility menu, dialog, popup

UserName

CAS RN 927802-38-8

H3CONHN

CAS RN 927802-38-8

IUPAC Name: 1-(2-methoxyethyl)-1,4-diazepane
Synonyms: 1-(2-methoxyethyl)-1,4-diazepane
Molecular Weight (g/mol): 158.25
Molecular Formula: C8H18N2O
InChi Key: PDGPTNXEAQRUMD-UHFFFAOYSA-N
SMILES: COCCN1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results