CAS RN 927961-18-0
CAS RN 927961-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lanifibranor, MedChemExpress
MedChemExpress Lanifibranor is a pan peroxisome proliferator-activated receptor (PPAR) agonist with EC50s of 1.5, 0.87 and 0.21 μM for human PPARα, PPARσ and PPARγ, respectively.