CAS RN 929-05-5
IUPAC Name:
1-ethoxybut-1-ene
Synonyms:
1-ethoxybut-1-ene
Molecular Weight (g/mol):
100.16
Molecular Formula:
C6H12O
InChi Key:
AQTYNINXYJFSHD-UHFFFAOYSA-N
SMILES:
CCOC=CCC
1
–
1
of
1
results
1
1-Butenyl ethyl ether, cis + trans, 98%
CAS: 929-05-5 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD03427069 InChI Key: AQTYNINXYJFSHD-AATRIKPKSA-N Synonym: e-1-ethoxybut-1-ene,1-butenyl ethyl ether,1e-1-ethoxybut-1-ene,1-ethoxybut-1-ene,1-ethoxy-1-butene,cis-1-butenyl ethyl ether,trans-1-butenyl ethyl ether,1-butene, 1-ethoxy-, e,trans-1-ethoxy-1-butene,1e-1-butenyl ethyl ether # PubChem CID: 5364930 IUPAC Name: (E)-1-ethoxybut-1-ene SMILES: CCC=COCC