accessibility menu, dialog, popup

UserName

CH3NCH3NH2

CAS RN 93-05-0

IUPAC Name: N1,N1-diethylbenzene-1,4-diamine
Synonyms: N,N-diethyl-1,4-benzenediamine
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: QNGVNLMMEQUVQK-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(N)C=C1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)