accessibility menu, dialog, popup

UserName

CAS RN 93-10-7

CAS RN 93-10-7

IUPAC Name: quinoline-2-carboxylic acid
Synonyms: quinaldic acid 2-chinolincarbonsaeure 2-quinolinylcarboxylate quinaldinate chinaldinsaeure quinaldinic acid 2-quinolinylcarboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: LOAUVZALPPNFOQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=CC=CC2=N1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)