accessibility menu, dialog, popup

UserName

CAS RN 93-85-6

H2NSNOHO

CAS RN 93-85-6

IUPAC Name: 2-amino-1,3-benzothiazole-6-carboxylic acid
Synonyms: 2-amino-1,3-benzothiazole-6-carboxylic acid
Molecular Weight (g/mol): 194.21
Molecular Formula: C8H6N2O2S
InChi Key: ZEAKWWWXCZMODH-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=C(C=C2S1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)