accessibility menu, dialog, popup

UserName

CAS RN 930-21-2

OHN

CAS RN 930-21-2

IUPAC Name: azetidin-2-one
Synonyms: 2-azetidinone propiolactam β-propiolactam
Molecular Weight (g/mol): 71.08
Molecular Formula: C3H5NO
InChi Key: MNFORVFSTILPAW-UHFFFAOYSA-N
SMILES: O=C1CCN1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results