accessibility menu, dialog, popup

UserName

CH3ONSiCH3H3CH3C

CAS RN 93102-05-7

IUPAC Name: benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine
Synonyms: benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine
Molecular Weight (g/mol): 237.42
Molecular Formula: C13H23NOSi
InChi Key: RPZAAFUKDPKTKP-UHFFFAOYSA-N
SMILES: COCN(CC1=CC=CC=C1)C[Si](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)