accessibility menu, dialog, popup

UserName

CAS RN 93139-85-6

H3COOBrOOCH3

CAS RN 93139-85-6

IUPAC Name: 1,3-diethyl 2-(4-bromophenyl)propanedioate
Synonyms: 1,3-diethyl 2-(4-bromophenyl)propanedioate
Molecular Weight (g/mol): 315.16
Molecular Formula: C13H15BrO4
InChi Key: NEEVEYWBDFTMKS-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(=O)OCC)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results