CAS RN 932-32-1
IUPAC Name:
2-chloro-N-methylaniline
Synonyms:
2-chloro-N-methylaniline
Molecular Weight (g/mol):
141.6
Molecular Formula:
C7H8ClN
InChi Key:
WGNNILPYHCKCFF-UHFFFAOYSA-N
SMILES:
CNC1=CC=CC=C1Cl
1
–
1
of
1
results
1
2-Chloro-N-methylaniline, 97%
CAS: 932-32-1 Molecular Formula: C7H8ClN Molecular Weight (g/mol): 141.598 MDL Number: MFCD00045170 InChI Key: WGNNILPYHCKCFF-UHFFFAOYSA-N Synonym: n-methyl-2-chloroaniline,benzenamine, 2-chloro-n-methyl,n1-methyl-2-chloroaniline,2-chloro-n-methylbenzenamine,o-chlormonomethylanilin,2-chlor-n-methylaniline,2-chloro-n-methyl aniline,2-chloro-n-methyl-aniline,2-methylamino chlorobenzene,2-chloranyl-n-methyl-aniline PubChem CID: 136736 IUPAC Name: 2-chloro-N-methylaniline SMILES: CNC1=CC=CC=C1Cl