accessibility menu, dialog, popup

UserName

NH2NHOHNO

CAS RN 932-52-5

IUPAC Name: 5-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-amino-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: BISHACNKZIBDFM-UHFFFAOYSA-N
SMILES: NC1=CNC(=O)NC1=O
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)