CAS RN 932033-57-3
CAS RN 932033-57-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
2,2’-Bis-(1-adamantyl)-4,4’-dimethoxybiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2’-Bis-(1-adamantyl)-4,4’-dimethoxybiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2'-Bis-(1-adamantyl)-4,4'-dimethoxybiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
1,1'-[4,4'Bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.