accessibility menu, dialog, popup

UserName

CAS RN 934389-88-5

ONOOHCl

CAS RN 934389-88-5

IUPAC Name: 2-(morpholin-4-yl)-8-phenyl-4H-chromen-4-one hydrochloride
Synonyms: 2-(morpholin-4-yl)-8-phenylchromen-4-one hydrochloride
Molecular Weight (g/mol): 343.81
Molecular Formula: C19H18ClNO3
InChi Key: OQZQSRICUOWBLW-UHFFFAOYSA-N
SMILES: Cl.O=C1C=C(OC2=C(C=CC=C12)C1=CC=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results